Assay Details
| Gene Symbol: LOC4339860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785519.2 | XP_015641005.1 | 10 - 11 | 1767 | 117 | |
| XM_015785520.2 | XP_015641006.1 | 10 - 11 | 1750 | 117 | ||
| XM_015785521.2 | XP_015641007.1 | 10 - 11 | 1747 | 117 | ||
| GenBank mRNA | AK099594.1 | - | 10 - 11 | 1769 | 117 | |
More Information
Set Membership: |
|

LOC4339860
Genomic Map