Assay Details
| Gene Symbol: LOC4339860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785519.2 | XP_015641005.1 | 12 - 13 | 1997 | 73 | |
| XM_015785520.2 | XP_015641006.1 | 12 - 13 | 1980 | 73 | ||
| XM_015785521.2 | XP_015641007.1 | 12 - 13 | 1977 | 73 | ||
| GenBank mRNA | AK099594.1 | - | 12 - 13 | 1999 | 73 | |
More Information
Set Membership: |
|

LOC4339860
Genomic Map