Assay Details
| Gene Symbol: LOC4339860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785519.2 | XP_015641005.1 | 1 - 2 | 383 | 104 | |
| XM_015785520.2 | XP_015641006.1 | 1 - 2 | 384 | 104 | ||
| XM_015785521.2 | XP_015641007.1 | 1 - 2 | 384 | 104 | ||
| GenBank mRNA | AK099594.1 | - | 1 - 2 | 387 | 104 | |
More Information
Set Membership: |
|

LOC4339860
Genomic Map