Assay Details
| Gene Symbol: LOC4339860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785519.2 | XP_015641005.1 | 8 - 9 | 1611 | 124 | |
| XM_015785520.2 | XP_015641006.1 | 8 - 9 | 1594 | 124 | ||
| XM_015785521.2 | XP_015641007.1 | 8 - 9 | 1591 | 124 | ||
| GenBank mRNA | AK099594.1 | - | 8 - 9 | 1615 | 122 | |
More Information
Set Membership: |
|

LOC4339860
Genomic Map