Assay Details
| Gene Symbol: LOC4339908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786477.2 | XP_015641963.1 | 1 | 247 | 74 | |
| GenBank mRNA | AY600991.1 | - | 1 | 68 | 74 | |
More Information
Set Membership: |
|

LOC4339908
Genomic Map