Assay Details
| Gene Symbol: LOC4339967 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK069048.1 | - | 2 - 3 | 908 | 114 | |
| AK099098.1 | - | 2 - 3 | 907 | 114 | ||
| AK100438.1 | - | 3 - 4 | 823 | 114 | ||
More Information
Set Membership: |
|

LOC4339967
Genomic Map