Assay Details
| Gene Symbol: LOC4339967 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK060795.1 | - | 3 - 4 | 232 | 139 | |
| AK069048.1 | - | 4 - 5 | 1088 | 139 | ||
| AK099098.1 | - | 4 - 5 | 1087 | 139 | ||
| AK100438.1 | - | 5 - 6 | 1003 | 139 | ||
More Information
Set Membership: |
|

LOC4339967
Genomic Map