Assay Details
| Gene Symbol: LOC4340000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785764.2 | XP_015641250.1 | 10 - 11 | 1166 | 65 | |
| GenBank mRNA | AY332474.1 | - | 10 - 11 | 934 | 65 | |
| GQ423058.1 | - | 10 - 11 | 878 | 65 | ||
More Information
Set Membership: |
|

LOC4340000
Genomic Map