Assay Details
| Gene Symbol: LOC4340000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785764.2 | XP_015641250.1 | 13 - 14 | 1388 | 70 | |
| GenBank mRNA | AY332474.1 | - | 13 - 14 | 1156 | 70 | |
More Information
Set Membership: |
|

LOC4340000
Genomic Map