Assay Details
| Gene Symbol: LOC4340000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785764.2 | XP_015641250.1 | 16 - 17 | 1603 | 71 | |
| GenBank mRNA | AY332474.1 | - | 16 - 17 | 1371 | 71 | |
More Information
Set Membership: |
|

LOC4340000
Genomic Map