Assay Details
| Gene Symbol: LOC4340000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785764.2 | XP_015641250.1 | 22 - 23 | 2032 | 118 | |
| GenBank mRNA | AK064052.1 | - | 3 - 4 | 224 | 118 | |
| AY332474.1 | - | 22 - 23 | 1857 | 118 | ||
| GQ423058.1 | - | 22 - 23 | 1744 | 118 | ||
More Information
Set Membership: |
|

LOC4340000
Genomic Map