Assay Details
| Gene Symbol: LOC4340000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785764.2 | XP_015641250.1 | 23 - 24 | 2168 | 67 | |
| GenBank mRNA | AK064052.1 | - | 4 - 5 | 360 | 67 | |
| AY332474.1 | - | 23 - 24 | 1993 | 67 | ||
| GQ423058.1 | - | 23 - 24 | 1880 | 67 | ||
More Information
Set Membership: |
|

LOC4340000
Genomic Map