Assay Details
| Gene Symbol: LOC4340000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785764.2 | XP_015641250.1 | 24 - 25 | 2513 | 99 | |
| GenBank mRNA | AK064052.1 | - | 5 - 6 | 705 | 99 | |
| AY332474.1 | - | 24 - 25 | 2338 | 99 | ||
More Information
Set Membership: |
|

LOC4340000
Genomic Map