Assay Details
| Gene Symbol: LOC4340000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785764.2 | XP_015641250.1 | 25 - 26 | 2878 | 69 | |
| GenBank mRNA | AK064052.1 | - | 6 - 7 | 1070 | 69 | |
| AY332474.1 | - | 25 - 26 | 2703 | 69 | ||
| GQ423058.1 | - | 25 - 26 | 2590 | 69 | ||
More Information
Set Membership: |
|

LOC4340000
Genomic Map