Assay Details
| Gene Symbol: LOC4340008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787552.2 | XP_015643038.1 | 1 | 876 | 92 | |
| GenBank mRNA | EU220429.2 | - | 1 | 864 | 92 | |
More Information
Set Membership: |
|

LOC4340008
Genomic Map