Assay Details
| Gene Symbol: LOC4340218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786410.2 | XP_015641896.1 | 1 - 2 | 413 | 58 | |
| GenBank mRNA | AK064184.1 | - | 1 - 2 | 320 | 58 | |
| U78890.1 | - | 1 - 2 | 307 | 58 | ||
More Information
Set Membership: |
|

LOC4340218
Genomic Map