Assay Details
| Gene Symbol: LOC4340218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786410.2 | XP_015641896.1 | 2 - 3 | 503 | 113 | |
| GenBank mRNA | AK064184.1 | - | 2 - 3 | 410 | 113 | |
| U78890.1 | - | 2 - 3 | 397 | 113 | ||
More Information
Set Membership: |
|

LOC4340218
Genomic Map