Assay Details
| Gene Symbol: LOC4340350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787233.2 | XP_015642719.1 | 1 - 2 | 982 | 78 | |
| GenBank mRNA | AK106963.1 | - | 1 - 2 | 551 | 78 | |
More Information
Set Membership: |
|

LOC4340350
Genomic Map