Assay Details
| Gene Symbol: LOC4340354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787927.2 | XP_015643413.1 | 6 - 7 | 1696 | 82 | |
| GenBank mRNA | AK066305.1 | - | 1 - 2 | 85 | 82 | |
More Information
Set Membership: |
|

LOC4340354
Genomic Map