Assay Details
| Gene Symbol: LOC4340925 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788408.2 | XP_015643894.1 | 4 - 5 | 550 | 84 | |
| GenBank mRNA | AK058294.1 | - | 4 - 5 | 555 | 84 | |
| AK101738.1 | - | 4 - 5 | 531 | 84 | ||
More Information
Set Membership: |
|

LOC4340925
Genomic Map