Assay Details
| Gene Symbol: LOC4340925 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788408.2 | XP_015643894.1 | 5 - 6 | 719 | 87 | |
| GenBank mRNA | AK058294.1 | - | 5 - 6 | 724 | 87 | |
| AK101738.1 | - | 5 - 6 | 700 | 87 | ||
More Information
Set Membership: |
|

LOC4340925
Genomic Map