Assay Details
| Gene Symbol: LOC4340947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789023.2 | XP_015644509.1 | 8 - 9 | 818 | 115 | |
| GenBank mRNA | AK100837.1 | - | 8 - 9 | 808 | 115 | |
More Information
Set Membership: |
|

LOC4340947
Genomic Map