Assay Details
| Gene Symbol: LOC4326459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766332.1 | XP_015621818.1 | 13 - 14 | 1569 | 117 | |
| GenBank mRNA | AK243573.1 | - | 13 - 14 | 1533 | 117 | |
| AY972084.1 | - | 13 - 14 | 1412 | 117 | ||
| BT132384.1 | - | 1 - 2 | 144 | 117 | ||
| EU152208.1 | - | 12 - 13 | 1679 | 117 | ||
More Information
Set Membership: |
|

LOC4326459
Genomic Map