Assay Details
| Gene Symbol: LOC9272141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766350.2 | XP_015621836.1 | 13 - 14 | 1256 | 69 | |
| XM_015766351.2 | XP_015621837.1 | 13 - 14 | 1256 | 69 | ||
| GenBank mRNA | AK242985.1 | - | 13 - 14 | 1047 | 69 | |
More Information
Set Membership: |
|

LOC9272141
Genomic Map