Assay Details
| Gene Symbol: LOC4327647 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791123.2 | XP_015646609.1 | 1 - 2 | 216 | 63 | |
| GenBank mRNA | AK100365.1 | - | 1 - 2 | 146 | 63 | |
| AK119523.1 | - | 1 - 2 | 83 | 63 | ||
| D73411.1 | - | 1 - 2 | 145 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015791123.2 transcripts.
Set Membership: |
|

LOC4327647
Genomic Map