Assay Details
| Gene Symbol: LOC4327311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770120.2 | XP_015625606.1 | 2 - 3 | 824 | 86 | |
| XM_015770128.2 | XP_015625614.1 | 2 - 3 | 824 | 86 | ||
| GenBank mRNA | AK068784.1 | - | 2 - 3 | 814 | 86 | |
More Information
Set Membership: |
|

LOC4327311
Genomic Map