Assay Details
| Gene Symbol: LOC4327326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767688.2 | XP_015623174.1 | 1 | 476 | 59 | |
| XM_015767695.2 | XP_015623181.1 | 1 | 486 | 59 | ||
| GenBank mRNA | AK243372.1 | - | 1 | 432 | 59 | |
More Information
Set Membership: |
|

LOC4327326
Genomic Map