Assay Details
| Gene Symbol: LOC4325116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766514.2 | XP_015622000.1 | 3 - 4 | 640 | 67 | |
| GenBank mRNA | AK243649.1 | - | 3 - 4 | 599 | 67 | |
More Information
Set Membership: |
|

LOC4325116
Genomic Map