Assay Details
| Gene Symbol: LOC4324441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766626.2 | XP_015622112.1 | 1 - 2 | 128 | 103 | |
| GenBank mRNA | AK058917.1 | - | 1 - 2 | 107 | 103 | |
| AK119715.1 | - | 1 - 2 | 104 | 103 | ||
| AK122128.1 | - | 1 - 2 | 104 | 103 | ||
| BT132259.1 | - | 1 - 2 | 96 | 103 | ||
| BT132504.1 | - | 1 - 2 | 103 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015766626.2 transcripts.
Set Membership: |
|

LOC4324441
Genomic Map