Assay Details
| Gene Symbol: LOC4324441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766626.2 | XP_015622112.1 | 2 - 3 | 234 | 83 | |
| GenBank mRNA | AK058917.1 | - | 2 - 3 | 213 | 83 | |
| AK119715.1 | - | 2 - 3 | 210 | 83 | ||
| AK122128.1 | - | 2 - 3 | 210 | 83 | ||
| BT132504.1 | - | 2 - 3 | 209 | 83 | ||
More Information
Set Membership: |
|

LOC4324441
Genomic Map