Assay Details
| Gene Symbol: LOC4326726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775332.2 | XP_015630818.1 | 1 | 662 | 119 | |
| GenBank mRNA | AK241615.1 | - | 1 | 646 | 119 | |
More Information
Set Membership: |
|

LOC4326726
Genomic Map