Assay Details
| Gene Symbol: LOC4326030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791944.2 | XP_015647430.1 | 12 - 13 | 1921 | 113 | |
| XM_015791954.2 | XP_015647440.1 | 11 - 12 | 1840 | 113 | ||
| GenBank mRNA | AK065466.1 | - | 12 - 13 | 1814 | 113 | |
More Information
Set Membership: |
|

LOC4326030
Genomic Map