Assay Details
| Gene Symbol: LOC4326030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791944.2 | XP_015647430.1 | 5 - 6 | 875 | 88 | |
| XM_015791954.2 | XP_015647440.1 | 4 - 5 | 794 | 88 | ||
| GenBank mRNA | AK065466.1 | - | 5 - 6 | 768 | 88 | |
More Information
Set Membership: |
|

LOC4326030
Genomic Map