Assay Details
| Gene Symbol: LOC4324047 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760852.1 | XP_015616338.1 | 1 - 2 | 240 | 77 | |
| XM_015760857.2 | XP_015616343.1 | 1 - 2 | 202 | 77 | ||
| GenBank mRNA | AK059119.1 | - | 1 - 2 | 208 | 77 | |
| AK102891.1 | - | 1 - 2 | 208 | 77 | ||
More Information
Set Membership: |
|

LOC4324047
Genomic Map