Assay Details
| Gene Symbol: LOC4326337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766433.1 | XP_015621919.1 | 8 - 9 | 987 | 86 | |
| GenBank mRNA | AK100223.1 | - | 8 - 9 | 965 | 86 | |
| JX134099.1 | - | 8 - 9 | 890 | 86 | ||
More Information
Set Membership: |
|

LOC4326337
Genomic Map