Assay Details
| Gene Symbol: LOC4326378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785466.2 | XP_015640952.1 | 4 - 5 | 746 | 77 | |
| GenBank mRNA | AK099544.1 | - | 4 - 5 | 705 | 77 | |
| AK102072.1 | - | 4 - 5 | 743 | 77 | ||
More Information
Set Membership: |
|

LOC4326378
Genomic Map