Assay Details
| Gene Symbol: LOC4326378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785466.2 | XP_015640952.1 | 6 - 7 | 1055 | 129 | |
| GenBank mRNA | AK099544.1 | - | 6 - 7 | 1012 | 129 | |
More Information
Set Membership: |
|

LOC4326378
Genomic Map