Assay Details
| Gene Symbol: LOC4327727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783110.2 | XP_015638596.1 | 6 - 7 | 1187 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4327727| Gene Symbol: LOC4327727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783110.2 | XP_015638596.1 | 6 - 7 | 1187 | 95 | |
Set Membership: |
|