Assay Details
| Gene Symbol: LOC4326639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795340.2 | XP_015650826.1 | 6 - 7 | 1157 | 61 | |
| GenBank mRNA | AK064316.1 | - | 6 - 7 | 997 | 61 | |
| AK102399.1 | - | 6 - 7 | 1081 | 61 | ||
More Information
Set Membership: |
|

LOC4326639
Genomic Map