Assay Details
| Gene Symbol: LOC4326570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767009.2 | XP_015622495.1 | 5 - 6 | 648 | 111 | |
| GenBank mRNA | AK102655.1 | - | 5 - 6 | 607 | 111 | |
| AK111923.1 | - | 5 - 6 | 623 | 111 | ||
More Information
Set Membership: |
|

LOC4326570
Genomic Map