Assay Details
| Gene Symbol: LOC4326570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767009.2 | XP_015622495.1 | 6 - 7 | 728 | 67 | |
| GenBank mRNA | AK102655.1 | - | 6 - 7 | 687 | 67 | |
| AK111923.1 | - | 6 - 7 | 703 | 67 | ||
More Information
Set Membership: |
|

LOC4326570
Genomic Map