Assay Details
| Gene Symbol: LOC4327858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766154.2 | XP_015621640.1 | 2 - 3 | 479 | 75 | |
| GenBank mRNA | AK062234.1 | - | 2 - 3 | 476 | 75 | |
| AK098887.1 | - | 2 - 3 | 416 | 75 | ||
| AK119723.1 | - | 2 - 3 | 426 | 75 | ||
More Information
Set Membership: |
|

LOC4327858
Genomic Map