Assay Details
| Gene Symbol: LOC4327862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759452.2 | XP_015614938.1 | 6 - 7 | 589 | 95 | |
| GenBank mRNA | AK058452.1 | - | 6 - 7 | 595 | 95 | |
| AK060072.1 | - | 6 - 7 | 583 | 95 | ||
More Information
Set Membership: |
|

LOC4327862
Genomic Map