Assay Details
| Gene Symbol: LOC4325515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766172.2 | XP_015621658.1 | 1 - 2 | 437 | 108 | |
| GenBank mRNA | AK061495.1 | - | 1 - 2 | 362 | 108 | |
| AK102396.1 | - | 1 - 2 | 351 | 108 | ||
| AK106121.1 | - | 1 - 2 | 436 | 108 | ||
| AK287679.1 | - | 384 | 108 | |||
More Information
Set Membership: |
|

LOC4325515
Genomic Map