Assay Details
| Gene Symbol: LOC4326924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763910.2 | XP_015619396.1 | 1 - 2 | 130 | 68 | |
| XM_015763925.2 | XP_015619411.1 | 1 - 2 | 130 | 68 | ||
| GenBank mRNA | AK100961.1 | - | 1 - 2 | 57 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015763910.2, XM_015763925.2 transcripts.
Set Membership: |
|

LOC4326924
Genomic Map