Assay Details
| Gene Symbol: LOC4326924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763901.2 | XP_015619387.1 | 7 - 8 | 2606 | 118 | |
| XM_015763910.2 | XP_015619396.1 | 8 - 9 | 2634 | 118 | ||
| XM_015763925.2 | XP_015619411.1 | 8 - 9 | 2616 | 118 | ||
| GenBank mRNA | AK100961.1 | - | 8 - 9 | 2543 | 118 | |
More Information
Set Membership: |
|

LOC4326924
Genomic Map