Assay Details
| Gene Symbol: LOC4325141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765392.2 | XP_015620878.1 | 8 - 9 | 1090 | 124 | |
| GenBank mRNA | AK105125.1 | - | 1 - 2 | 214 | 124 | |
More Information
Set Membership: |
|

LOC4325141
Genomic Map