Assay Details
| Gene Symbol: LOC4325622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788973.2 | XP_015644459.1 | 2 - 3 | 602 | 72 | |
| GenBank mRNA | AB101652.1 | - | 1 - 2 | 47 | 72 | |
More Information
Set Membership: |
|

LOC4325622
Genomic Map