Assay Details
| Gene Symbol: LOC4324711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765311.2 | XP_015620797.1 | 12 - 13 | 1317 | 91 | |
| XM_015765321.2 | 8 - 9 | 572 | 91 | |||
More Information
Set Membership: |
|

LOC4324711
Genomic Map