Assay Details
| Gene Symbol: LOC4324711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765311.2 | XP_015620797.1 | 21 - 22 | 2097 | 114 | |
| XM_015765321.2 | 17 - 18 | 1352 | 114 | |||
More Information
Set Membership: |
|

LOC4324711
Genomic Map